2,2,4-Trimethylpentane, extra dry 99.5%, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
326940010
326941000
TS326940010EA
120
USD
TS326940010
TS326941000
2,2,4-Trimethylpentane, extra dry 99.5%, AcroSeal®
2,2,4-Trimethylpentane
Formula:
(CH₃)₂CHCH₂C(CH₃)₃ Boiling Pt: 98…99 °C (760 mmHg) Melting Pt: –107 °C Density: 0.69 g/cm³ Flash Pt: –12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008943 CAS Number: 540-84-1 EINECS: 208-759-1 UN: 1262 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.4 % |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...