4-Methyl-2-pentanone 99.5%, Spectrophotometric Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
222170010
222170025
222175000
TS222170010EA
97
USD
TS222170010
TS222170025
TS222175000
4-Methyl-2-pentanone 99.5%, Spectrophotometric Grade
4-Methyl-2-pentanone
Formula:
(CH₃)₂CHCH₂COCH₃ Boiling Pt: 117.4 °C (760 mmHg) Melting Pt: –84 °C Density: 0.8 g/cm³ Flash Pt: 14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.05 % |
GC | ≥99.5 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤15 APHA |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...