4-Methyl-2-pentanone 98.5+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
423960025
423965000
423960010
TS423960025EA
156
USD
TS423960025
TS423965000
TS423960010
4-Methyl-2-pentanone 98.5+% ACS
4-Methyl-2-pentanone
Formula:
(CH₃)₂CHCH₂COCH₃ Boiling Pt: 117.4 °C (1013 hPa) Melting Pt: –84 °C Density: 0.8 g/cm³ Flash Pt: 14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3950 to 1.3970 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Residue after evaporation | ≤0.005 % |
Titrable acid | ≤0.002 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...