n-Pentane, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentane

397230010 397231000
TS397230010EA 122 USD
TS397230010 TS397231000
n-Pentane, extra dry 99+%, AcroSeal™
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3 II
Merck Index: 13,07193

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR