n-Pentane, anhydrous 99+%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentane
217240010
217240025
217240251
TS217240010EA
114
USD
TS217240010
TS217240025
TS217240251
n-Pentane, anhydrous 99+%, Extra Pure
n-Pentane
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36 °C (760 mmHg) Melting Pt: –130 °C Density: 0.626 g/cm³ Flash Pt: –49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II REACH: 01-2119459286-30 Merck Index: 13,07193 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3570 to 1.3590 (20°C, 589 nm) |
Water | ≤0.005 % |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...