n-Pentane 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentane

389070010 389070025
TS389070010EA 115 USD
TS389070010 TS389070025
n-Pentane 99+% for HPLC
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 %
GC ≥99.0 %
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR