n-Dodecane, anhydrous 99%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

434590010 434591000
TS434590010EA 320 USD
TS434590010 TS434591000
n-Dodecane, anhydrous 99%, AcroSeal®
n-Dodecane
Formula: H₃C(CH₂)₁₀CH₃
Boiling Pt: 215…217 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0.753 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR