Glycerine 83,5-88,5 w/w aqueous solution, pure

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

277360010 277360050
TS277360010EA 105 USD
TS277360010 TS277360050
Glycerine 83,5-88,5 w/w aqueous solution, pure
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.227 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497

Order Now

Specification Test Results

Appearance Clear colorless to almost colorless Viscous liquid
Refractive index 1.4490 to 1.4550 (20°C, 589 nm)
Assay 83.5 to 88.5 %
Water 12 to 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR