1-Propanol, extra dry 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

396940010 396941000
TS396940010EA 118 USD
TS396940010 TS396941000
1-Propanol, extra dry 99.5%, AcroSeal™
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –127 °C
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
UN: 1274
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.5 %
Residue after evaporation ≤0.001 %
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR