Nitromethane 96% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
445670010
445670025
445671000
TS445670010EA
145
USD
TS445670010
TS445670025
TS445671000
Nitromethane 96% for HPLC
Nitromethane
Formula:
CH₃NO₂ Boiling Pt: 101.2 °C (1013 hPa) Melting Pt: –29 °C Density: 1.12 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007400 CAS Number: 75-52-5 UN: 1261 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.03 % |
GC | ≥95 % |
UV | at 430 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...