Chloroform 99+% Stabilised analysis_for

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: EtOH

232090010 232090025 232090250
TS232090010EA 88 USD
TS232090010 TS232090025 TS232090250
Chloroform 99+% Stabilised analysis_for
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.01 %
GC ≥99.0 % (Excluding stabilizer)
Color scale ≤10 APHA
Stabilizer 0.6 to 1.0 % ethanol
Residue after evaporation ≤0.001 %
Free chlorine Passes test
Cadmium (Cd) ≤0.1 ppm
Cobalt (Co) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.5 ppm
Nickel (Ni) ≤0.1 ppm
Zinc (Zn) ≤0.5 ppm
Acidity (HCl) ≤0.001 %
Acetone and aldehyde ≤0.005 % (as Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR