Chloroform 99+% Stabilised, extra_pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Amylen

383760010 383760025 383760250
TS383760010EA 86 USD
TS383760010 TS383760025 TS383760250
Chloroform 99+% Stabilised, extra_pure
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer Approx. 100 ppm Amylene
Residue after evaporation ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR