1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

443010010 443011000
TS443010010EA 107 USD
TS443010010 TS443011000
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
Boiling Pt: 81…85 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.25 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
UN: 1184
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR