1,2-Dichloroethane 99+% ACS, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals
Danger

406820010 406820025 406825000
TS406820010EA 117 USD
TS406820010 TS406820025 TS406825000
1,2-Dichloroethane 99+% ACS, meets analytical specifications of Reagent Ph. Eur.
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
Boiling Pt: 81…85 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.25 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (Coulometric)
Density (20/20°C) about 1.25
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Titrable acid ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR