1,2-Dichloroethane 99+%, pure

Supplier: Thermo Scientific Chemicals
Danger

406800010 406800025
TS406800010EA 58 USD
TS406800010 TS406800025
1,2-Dichloroethane 99+%, pure
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
Boiling Pt: 81…85 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.25 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR