Acetic acid ≥80% (v/v) in water

Supplier: Thermo Scientific Chemicals
Danger

456860050 456860250
TS456860050EA 58 USD
TS456860050 TS456860250
Acetic acid ≥80% (v/v) in water
Acetic acid
Formula: H₃CCOOH
Flash Pt: >60 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Density 1.067 to 1.071 kg/l
Titration with NaOH 79.0 to 81.0 % v/v


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR