Acetic acid 25% in water

Supplier: Thermo Scientific Chemicals
Danger

456850050 456850250
TS456850050EA 58 USD
TS456850050 TS456850250
Acetic acid 25% in water
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 103 °C (760 mmHg)
Melting Pt: –17 °C
Density: 1.04 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with NaOH 24.0 to 26.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR