Crystal Digital PCR® Assay for PIK3CA (H1047, H1047R)
Supplier: STILLA TECHNOLOGIES, INC.
Assay-specific Crystal Flex Probes and primers for the detection of PIK3CA mutations and its positive control. The PIK3CA (H1047, H1047R) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 1 mutation in the PIK3CA gene using the Ruby Chip. PIK3CA is essential for regulating multiple cellular processes through the PI3K/AKT/mTOR signaling pathway including cell growth, proliferation, survival, and metabolism.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Red, blue
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target ESR1: PIK3CA H1047/H1047R
Precaución: For research use only. Not for use in diagnostic procedures.