[Arg 8]-Vasopressin Acetate ≥98.0% (by HPLC)

Supplier: TCI America

V0159-25MG V0159-5MG
TCV0159-25MGEA 612.85 USD
TCV0159-25MG TCV0159-5MG
[Arg 8]-Vasopressin Acetate ≥98.0% (by HPLC)
[Arg 8]-Vasopressin Acetate
Formula: C₄₆H₆₅N₁₅O₁₂S₂·xC₂H₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00133939
CAS Number: 129979-57-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR