(±)-Sulfinpyrazone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1,2-Diphenyl-4-(2-(phenylsulphinyl)ethyl)pyrazolidine-3,5-dione, sulphinpyrazone, 1,2-Diphenyl-4-[2-(phenylsulphinyl)ethyl]pyrazolidine-3,5-dione

U0114-25G U0114-5G
TCU0114-25GEA 147.29 USD
TCU0114-25G TCU0114-5G
(±)-Sulfinpyrazone ≥98.0% (by HPLC, titration analysis)
(±)-Sulfinpyrazone
Formula: C₂₃H₂₀N₂O₃S
MW: 404.484 g/mol
MDL Number: MFCD00057279
CAS Number: 57-96-5
EINECS: 200-357-4
Merck Index: 14,08949
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e869447a01d28c0c5948b7a9acc8164497f4e58f&vid=881978f772d3e34bd1ccc5af2f4adce8ba85d2ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralisation titration) min. 95.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR