Triethoxy(2,4,4-trimethylpentyl)silane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3883-100ML T3883-25ML
TCT3883-100MLEA 91.67 USD
TCT3883-100ML TCT3883-25ML
Triethoxy(2,4,4-trimethylpentyl)silane ≥97.0% (by GC)
Triethoxy(2,4,4-trimethylpentyl)silane
Formula: C₁₄H₃₂O₃Si
Flash Pt: 42 °C
MDL Number: MFCD09838917
CAS Number: 35435-21-3
EINECS: 252-558-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR