2-(2,3,4-Trifluorophenyl)pyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T3868-1G T3868-200MG
TCT3868-1GEA 174.07 USD
TCT3868-1G TCT3868-200MG
2-(2,3,4-Trifluorophenyl)pyridine ≥97.0% (by GC, titration analysis)
2-(2,3,4-Trifluorophenyl)pyridine
Formula: C₁₁H₆F₃N
MW: 209.17 g/mol
CAS Number: 1431374-74-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR