2,3,5-Trichlorophenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2,3,5-Trichlorobenzeneboronic acid, Dihydroxy(2,3,5-trichlorophenyl)borane, (2,3,5-Trichlorophenyl)metaboric acid

T3851-1G T3851-5G
TCT3851-1GEA 42.23 USD
TCT3851-1G TCT3851-5G
2,3,5-Trichlorophenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
2,3,5-Trichlorophenylboronic acid
Formula: C₆H₄BCl₃O₂
MW: 225.26 g/mol
MDL Number: MFCD01630856
CAS Number: 212779-19-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b195f6f7b1f66c71401a36ed68bf75172642eb34&vid=557ab62a5be7060c551b1cea3eeb95c654c2cd43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) 98.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR