4-(Trifluoromethyl)benzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 4-Aminomethylbenzotrifluoride hydrochloride, 4-(Trifluoromethyl)benzylammonium chloride, [4-(Trifluoromethyl)phenyl]methanamine hydrochloride

T3836-1G T3836-5G
TCT3836-1GEA 36.05 USD
TCT3836-1G TCT3836-5G
4-(Trifluoromethyl)benzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-(Trifluoromethyl)benzylamine hydrochloride
Formula: C₈H₈F₃N·HCl
Melting Pt: 167 °C
MDL Number: MFCD07780751
CAS Number: 3047-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=694863d7500e3c3123149e45c126c5bf755805b3&vid=439fd07d1bab91c4fc214cef738da17b69640dee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR