2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4,5,5-Tetramethyl-2-(1,1':3',1''-terphenyl)-3-yl-1,3,2-dioxaborolane, (1,1':3',1''-Terphenyl)-3-ylboronic acid Pinacol ester

T3828-1G T3828-250MG
TCT3828-1GEA 99.91 USD
TCT3828-1G TCT3828-250MG
2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₂₄H₂₅BO₂
MW: 356.27 g/mol
MDL Number: MFCD28134531
CAS Number: 1115023-84-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR