Tasimelteon ≥98.0% (by HPLC)

Supplier: TCI America

T3813-10MG T3813-50MG
TCT3813-10MGEA 126.69 USD
TCT3813-10MG TCT3813-50MG
Tasimelteon ≥98.0% (by HPLC)
Tasimelteon
Formula: C₁₅H₁₉NO₂
Melting Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09033789
CAS Number: 609799-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa499d5c44c3846901d62a0c7394e44446014134&vid=1c6580dbf21a7044481e0505dd25cb14c578c764&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.30 to 6.10 %
Specific rotation [a]20/D -41.0 to -46.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR