4-[1,2,4]Triazol-4-ylbenzoic acid ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4-(4H-1,2,4-Triazol-4-yl)benzoic acid, 4-(p-Carboxyphenyl)-1,2,4-triazole

T3788-1G T3788-200MG
TCT3788-1GEA 280.16 USD
TCT3788-1G TCT3788-200MG
4-[1,2,4]Triazol-4-ylbenzoic acid ≥98.0% (by HPLC)
4-[1,2,4]Triazol-4-ylbenzoic acid
Formula: C₉H₇N₃O₂
MW: 189.18 g/mol
MDL Number: MFCD06654839
CAS Number: 157069-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7422662a399c03444a8582a08458ba271cccd69&vid=88b7a7fbaf1854c5ad2bb1fe7f265125fc6964e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR