4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Trifluoromethylphenyl)benzaldehyde, 4'-Trifluoromethyl-biphenyl-4-carbaldehyde, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

T3755-1G T3755-5G
TCT3755-1GEA 45.32 USD
TCT3755-1G TCT3755-5G
4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥98.0% (by GC)
4'-Trifluoromethylbiphenyl-4-carbaldehyde
Formula: C₁₄H₉F₃O
MW: 250.2200 g/mol
MDL Number: MFCD01862519
CAS Number: 90035-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10d527cc6faca343e0df4017084999456407ddf6&vid=5211a00e57a3b87b2eb7696242f92377b58ab210&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR