Tectoridin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 7-(beta-D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 5-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Tectorigenin 7-O-beta-D-Glucopyranoside

T3724-10MG T3724-50MG
TCT3724-10MGEA 98.88 USD
TCT3724-10MG TCT3724-50MG
Tectoridin ≥98.0% (by HPLC)
Tectoridin
Formula: C₂₂H₂₂O₁₁
MW: 462.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01662715
CAS Number: 611-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3142e6b8615f8fe15863f9b51b5422596d19805c&vid=97fcc010a7d8b404f994c00aeb458be3db6bb1c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR