Tris(ethyl Acetoacetato)aluminum(III) ≥97.0% (by titrimetric analysis) (contains ca. 2% 2-Butanol at maximum)

Supplier: TCI America

T3681-25G T3681-500G
TCT3681-25GEA 32.96 USD
TCT3681-25G TCT3681-500G
Tris(ethyl Acetoacetato)aluminum(III) ≥97.0% (by titrimetric analysis) (contains ca. 2% 2-Butanol at maximum)
Tris(ethyl Acetoacetato)aluminum(III)
Formula: C₁₈H₂₇AlO₉
MDL Number: MFCD01317537
CAS Number: 15306-17-9
EINECS: 239-343-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe732ee0869a3ee2f56904aba53dd4efa656675&vid=8a6e2955789d6aee3606812ea7178b6a64d91fbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR