2,2,2-Trifluoro-N-phenylacetamide ≥98.0% (by GC)

Supplier: TCI America

T3645-1G T3645-5G
TCT3645-1GEA 59.74 USD
TCT3645-1G TCT3645-5G
2,2,2-Trifluoro-N-phenylacetamide ≥98.0% (by GC)
2,2,2-Trifluoro-N-phenylacetamide
Formula: C₈H₆F₃NO
MDL Number: MFCD00196027
CAS Number: 404-24-0
EINECS: 206-965-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad265ca98bde33519a114d7f7e452421f24ada2&vid=b71fbcca4babf2e865dca4e11813f5e34ef7652c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR