(Heptafluoropropyl)trimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Trimethyl(heptafluoropropyl)silane, Trimethyl(perfluoropropyl)silane

T3593-1G T3593-5G
TCT3593-1GEA 86.52 USD
TCT3593-1G TCT3593-5G
(Heptafluoropropyl)trimethylsilane ≥98.0% (by GC)
(Heptafluoropropyl)trimethylsilane
Formula: C₆H₉F₇Si
Boiling Pt: 88 °C
Flash Pt: 54 °C
MDL Number: MFCD00216712
CAS Number: 3834-42-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37e73a3390d2167483f1fef3014faba1fa55fb1f&vid=ad08222d240e3e047303fceeca0a275031b44994&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR