Tin(II) bromide ≥97.0% (by titrimetric analysis) for perovskite precursor

Supplier: TCI America

Synonyms: Tin dibromide

T3573-1G T3573-5G
TCT3573-1GEA 60.77 USD
TCT3573-1G TCT3573-5G
Tin(II) bromide ≥97.0% (by titrimetric analysis) for perovskite precursor
Tin(II) bromide
Formula: Br₂Sn
MW: 278.52 g/mol
MDL Number: MFCD00011239
CAS Number: 10031-24-0
Merck Index: 12,08938
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7df3b4890d0e12b07d616533d1a00f9a2d5b1524&vid=fff6432e4f2c59c2e2d9bc020f41ccbf1f483b5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR