1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2',2''-(1,3,5-Benzenetriyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,3,5-Phenyltriboronic acid tris(pinacol) ester

T3515-1G T3515-5G
TCT3515-1GEA 89.61 USD
TCT3515-1G TCT3515-5G
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene ≥98.0% (by GC, titration analysis)
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Formula: C₂₄H₃₉B₃O₆
MW: 456.00 g/mol
MDL Number: MFCD17167305
CAS Number: 365564-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f779b4e4576e0b07a6ca26276203d0e38e43cd7&vid=a9a41f1208d8fa4e5af4383d8cc34fb263a6704a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR