Tetraethylammonium hydrogen sulphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Tetraethylammonium bisulphate, Tetraethylammonium hydrogensulphate

T3506-25G T3506-5G
TCT3506-25GEA 168.92 USD
TCT3506-25G TCT3506-5G
Tetraethylammonium hydrogen sulphate ≥98.0% (by titrimetric analysis)
Tetraethylammonium hydrogen sulphate
Formula: C₈H₂₁NO₄S
MW: 227.33
MDL Number: MFCD00036150
CAS Number: 16873-13-5
EINECS: 240-899-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78613a83e9dca6fc54c72f0ef0f73fed953e4be7&vid=425cf811aa25db81feb89d689ca191ef58c74526&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralisation titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR