4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)phenoxy]tetrahydro-2H-pyran, 4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-2-yl)oxy)phenyl)-1,3,2-dioxaborolane, 4-[(Tetrahydro-2H-pyran-2-yl)oxy]phenylboronic acid Pinacol ester

T3468-1G T3468-5G
TCT3468-1GEA 103 USD
TCT3468-1G TCT3468-5G
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane
Formula: C₁₇H₂₅BO₄
MW: 304.19 g/mol
MDL Number: MFCD06659834
CAS Number: 889865-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc171987d53ee8507e0566235339fce514d6a574&vid=3f0f8a7508afb5591a1ecd4a812f508446e64861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR