2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)

Supplier: TCI America

T3433-1G T3433-200MG
TCT3433-1GEA 369.77 USD
TCT3433-1G TCT3433-200MG
2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)
2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene]
Formula: C₇₅H₄₄
Melting Pt: 457 °C
MDL Number: MFCD12022449
CAS Number: 518997-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f65b6b5b2c8ee0d8ec72fccb7448621ddf60911&vid=31d36f796512b09cd72f090cc980e6335c4e6af4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 93.00 to 95.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR