2,2,2-Trifluoroethyl acetate ≥97.0% (by GC)
Supplier: TCI America
|
Danger
|
T3422-25G
T3422-5G
TCT3422-25GEA
162.74
USD
TCT3422-25G
TCT3422-5G
2,2,2-Trifluoroethyl acetate ≥97.0% (by GC)
2,2,2-Trifluoroethyl acetate
Formula:
C₄H₅F₃O₂ Boiling Pt: 78 °C Flash Pt: 11 °C |
MDL Number:
MFCD00198018 CAS Number: 406-95-1 UN: 3272 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...