2-Phenyl-2',4',6'-trihydroxyacetophenone ≥96.0% (by GC)

Supplier: TCI America
Warning

T3390-1G T3390-5G
TCT3390-1GEA 98.88 USD
TCT3390-1G TCT3390-5G
2-Phenyl-2',4',6'-trihydroxyacetophenone ≥96.0% (by GC)
2-Phenyl-2',4',6'-trihydroxyacetophenone
Formula: C₁₄H₁₂O₄
MDL Number: MFCD00205539
CAS Number: 727-71-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08cc2c41440567d2bdc72d01d523c384be6f9326&vid=f12676c440e370ff399fbb0a8e976bfc145a4ff1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR