2,2,2,4'-Tetrafluoroacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
T3257-25G
T3257-5G
TCT3257-25GEA
271.92
USD
TCT3257-25G
TCT3257-5G
2,2,2,4'-Tetrafluoroacetophenone ≥98.0% (by GC)
2,2,2,4'-Tetrafluoroacetophenone
Formula:
C₈H₄F₄O MW: 192.1104 g/mol Boiling Pt: 152 °C Flash Pt: 32 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042393 CAS Number: 655-32-3 EINECS: 211-509-4 UN: 1325 ADR: 4.1,II |
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...