2-(Trifluoromethyl)pyridine ≥95.0% (by GC)

Supplier: TCI America
Warning

T2683-1G T2683-5G
TCT2683-1GEA 20.6 USD
TCT2683-1G TCT2683-5G
2-(Trifluoromethyl)pyridine ≥95.0% (by GC)
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 140 °C
Flash Pt: 42 °C
MDL Number: MFCD02183562
CAS Number: 368-48-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7be7df871f3d54754616fa88a07877a0afb9e02&vid=b657b6607ee33c40bd491665189d4a2cc7c6c049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR