RS-127445 hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: RS-127445 HCl, 4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine hydrochloride

R0221-10MG R0221-50MG
TCR0221-10MGEA 217.33 USD
TCR0221-10MG TCR0221-50MG
RS-127445 hydrochloride ≥98.0% (by HPLC)
RS-127445 hydrochloride
Formula: C₁₇H₁₆FN₃·HCl
MW: 317.79 g/mol
Melting Pt: 158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11112196
CAS Number: 199864-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4444c0e736c7731843639ad30ef06d596fd3199&vid=7928d90a5438ae84e3dcc56416085b94c871fb2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 12.60 to 13.80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR