Sphynolactone-7 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: SPL 7, 4-Methyl-5-oxo-2,5-dihydrofuran-2-yl 4-[(4-Butoxyphenyl)sulphonyl]piperazine-1-carboxylate

P2745-25MG P2745-5MG
TCP2745-25MGEA 173.04 USD
TCP2745-25MG TCP2745-5MG
Sphynolactone-7 ≥98.0% (by HPLC)
Sphynolactone-7
Formula: C₂₀H₂₆N₂O₇S
MW: 438.50 g/mol
CAS Number: 2305752-57-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR