trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-ol, trans-4-(trans-4-Amylcyclohexyl)cyclohexanol, trans,trans-4'-Amyl-[1,1'-bi(cyclohexane)]-4-ol, trans,trans-4'-Pentyl-[1,1'-bi(cyclohexane)]-4-ol

P2690-25G P2690-5G
TCP2690-25GEA 320.33 USD
TCP2690-25G TCP2690-5G
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol ≥98.0% (by GC)
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol
Formula: C₁₇H₃₂O
MW: 252.44 g/mol
MDL Number: MFCD09752070
CAS Number: 82575-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7e8d674755ecea0a3798e05a544d3c79d69c4fb&vid=076be829d3c7056521e71a707f2fd476477c7c88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR