4-Phenoxybenzylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

P2664-25G P2664-5G
TCP2664-25GEA 193.64 USD
TCP2664-25G TCP2664-5G
4-Phenoxybenzylamine ≥98.0% (by GC)
4-Phenoxybenzylamine
Formula: C₁₃H₁₃NO
MDL Number: MFCD01310836
CAS Number: 107622-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51ac0d99533d146987f3024eacc3f22fd7e50b49&vid=75e1083d466bfe0c2f708d932035158a7de7cc85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR