Potassium (3-bromophenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: potassium (3-bromophenyl)trifluoroborate, (3-Bromophenyl)trifluoroboric acid potassium salt

P2646-1G P2646-5G
TCP2646-1GEA 57.68 USD
TCP2646-1G TCP2646-5G
Potassium (3-bromophenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)
Potassium (3-bromophenyl)trifluoroborate
Formula: C₆H₄BBrF₃K
MW: 262.91 g/mol
MDL Number: MFCD02093977
CAS Number: 374564-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b61ab39235900f2152d73ce33190421fd5545bf3&vid=1342cfc042c82d8473c3b5248790e701c384cde8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) min. 98.0 %
Melting point 186.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR