Propyl Levulinate ≥98.0% (by GC)

Supplier: TCI America

P2631-25G P2631-5G
TCP2631-25GEA 188.49 USD
TCP2631-25G TCP2631-5G
Propyl Levulinate ≥98.0% (by GC)
Propyl Levulinate
Formula: C₈H₁₄O₃
Boiling Pt: 221 °C
MDL Number: MFCD16041935
CAS Number: 645-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf9e3c1f884a1b0b22a4b34311f10cbb36611016&vid=da24801750aa6940fdbebe76fe41d0f45b38c8ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR