5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
5'-Phenyl-m-terphenyl-4-amine
P2589-1G
P2589-5G
TCP2589-1GEA
108.15
USD
TCP2589-1G
TCP2589-5G
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine ≥98.0% (by HPLC)
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine
Formula:
C₂₄H₁₉N MW: 321.42 g/mol |
MDL Number:
MFCD22581309 CAS Number: 343239-58-7 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 152.0 to 156.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...