PD 169316 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-(4-(4-Fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl)pyridine, 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine

P2532-10MG P2532-50MG
TCP2532-10MGEA 213.21 USD
TCP2532-10MG TCP2532-50MG
PD 169316 ≥98.0% (by HPLC)
PD 169316
Formula: C₂₀H₁₃FN₄O₂
MW: 360.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405019
CAS Number: 152121-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2648757315714ba85d0f1cff87016c01962ebda6&vid=8089b492b08140f6ca11260754b49eb7347a69b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.70 to 16.30 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR