2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,4-Diphenyl-6-(9,9'-spirobi[9H-fluoren]-2-yl)-1,3,5-triazine

P2512-1G P2512-200MG
TCP2512-1GEA 348.14 USD
TCP2512-1G TCP2512-200MG
2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC)
2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₄₀H₂₅N₃
MW: 547.66 g/mol
MDL Number: MFCD30721570
CAS Number: 1207176-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74af6e95b915f992520aed476f88fd86e6e45e30&vid=e7f839777ce7affbd1d1b6afceea292357d96c0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR